Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318810.1 | NP_001305739.1 | 2 | 566 | ||
NM_001318811.1 | NP_001305740.1 | 2 | 219 | |||
NM_014926.3 | NP_055741.2 | 2 | 404 | |||
GenBank mRNA | AB020655.1 | - | 2 | 233 | ||
AK290024.1 | - | 2 | 566 | |||
BC114621.1 | - | 2 | 218 | 40036558 | ||
DA123399.1 | - | 2 | 406 | |||
DA233757.1 | - | 2 | 219 |