Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001021.1 | NP_001001021.1 | 1 | 142 |
Gene Symbol: LOC100912515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229824.1 | XP_006229886.1 | 1 | 142 |