Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001511.2 | NP_001001511.1 | 1 | 1194 | ||
GenBank mRNA | AY435153.1 | - | 1 | 714 | ||
BC099796.1 | - | 1 | 1194 | 7936188 |