Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033117.2 | NP_001028289.1 | 22 | 7671 | ||
NM_014850.3 | NP_055665.1 | 22 | 7743 | |||
XM_011534295.1 | XP_011532597.1 | 23 | 7758 | |||
XM_011534296.1 | XP_011532598.1 | 24 | 7875 | |||
XM_011534301.2 | XP_011532603.1 | 20 | 9587 | |||
XM_011534303.1 | XP_011532605.1 | 18 | 6483 | |||
XM_017007574.1 | XP_016863063.1 | 22 | 8081 | |||
XM_017007575.1 | XP_016863064.1 | 22 | 8009 | |||
XM_017007576.1 | XP_016863065.1 | 24 | 7250 | |||
XM_017007577.1 | XP_016863066.1 | 24 | 7189 | |||
XM_017007578.1 | XP_016863067.1 | 22 | 8109 | |||
XM_017007579.1 | XP_016863068.1 | 21 | 7749 | |||
GenBank mRNA | AF427144.1 | - | 22 | 7111 | ||
AF464189.1 | - | 22 | 7199 |