Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001144.3 | NP_001001144.2 | 3 | 410 | ||
NM_001103162.2 | NP_001096632.1 | 3 | 322 | |||
GenBank mRNA | AK035432.1 | - | 3 | 301 | ||
AK047215.1 | - | 1 | 1249 | |||
AK146658.1 | - | 3 | 392 | |||
AK150228.1 | - | 2 | 326 | |||
AK150275.1 | - | 3 | 294 | |||
AK152478.1 | - | 3 | 294 | |||
AK153330.1 | - | 3 | 294 | |||
AK172912.1 | - | 3 | 301 | |||
BC070437.1 | - | 3 | 410 | 30535805 | ||
BC072633.1 | - | 3 | 283 | 6849461 | ||
BY015861.1 | - | 3 | 323 | |||
BY311416.1 | - | 3 | 323 |