Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001160.3 | NP_001001160.1 | 14 | 3768 | ||
NM_001289674.1 | NP_001276603.1 | 13 | 3599 | |||
NM_001289675.1 | NP_001276604.1 | 13 | 3593 | |||
GenBank mRNA | AK035911.1 | - | 2 | 703 | ||
AK129466.1 | - | 12 | 3145 | |||
AK147230.1 | - | 14 | 3660 | |||
AK147681.1 | - | 13 | 3458 | |||
BC061475.1 | - | 11 | 2240 | |||
BC070445.1 | - | 13 | 3599 | 30536738 |