Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001176.2 | NP_001001176.2 | 7 | 1231 | ||
NM_001167988.1 | NP_001161460.1 | 8 | 1169 | |||
GenBank mRNA | AK135097.1 | - | 7 | 1228 | ||
BC051635.1 | - | 7 | 1029 | |||
BC066223.1 | - | 7 | 1022 | 30468965 |