Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014904.2 | NP_055719.1 | 5 | 5190 | ||
XM_005269629.3 | XP_005269686.1 | 6 | 5568 | |||
GenBank mRNA | AB023158.1 | - | 5 | 5190 | ||
AY037299.1 | - | 5 | 5190 | |||
BX647717.1 | - | 4 | 3496 |