Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001496.2 | NP_001001496.1 | 2 | 1070 | ||
GenBank mRNA | AK040693.1 | - | 2 | 1030 | ||
AK146838.1 | - | 2 | 1070 | |||
AK168953.1 | - | 2 | 1070 | |||
BC117760.2 | - | 2 | 1013 | 40090837 |