Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014905.4 | NP_055720.3 | 14, 15 | 1921 | ||
XM_006712435.1 | XP_006712498.1 | 14, 15 | 1921 | |||
GenBank mRNA | AB020645.1 | - | 14, 15 | 1907 | ||
AF097493.1 | - | 7, 8 | 649 | |||
AF223943.1 | - | 14, 15 | 1676 | |||
AF327434.1 | - | 14, 15 | 1887 | |||
AK289747.1 | - | 15, 16 | 1897 | |||
AK294927.1 | - | 3, 4 | 445 | |||
AK298266.1 | - | 3, 4 | 273 | |||
BC038507.1 | - | 15, 16 | 1897 | 5263220 |