Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001552.4 | NP_001001552.3 | 2, 3 | 210 | ||
NM_001199050.1 | NP_001185979.1 | 2, 3 | 210 | |||
XM_011510160.2 | XP_011508462.1 | 2, 3 | 214 | |||
XM_011510161.2 | XP_011508463.1 | 2, 3 | 202 | |||
XM_011510162.2 | XP_011508464.1 | 2, 3 | 212 | |||
XM_011510164.2 | XP_011508466.1 | 2, 3 | 234 | |||
XM_011510165.2 | XP_011508467.1 | 2, 3 | 230 | |||
GenBank mRNA | AB085610.1 | - | 2, 3 | 177 | ||
AB096677.1 | - | 2, 3 | 177 | |||
AB096680.1 | - | 2, 3 | 177 | |||
AB096681.1 | - | 2, 3 | 177 | |||
AK026628.1 | - | 2, 3 | 177 | |||
AK302392.1 | - | 2, 3 | 212 | |||
BC036636.2 | - | 2, 3 | 180 | 5296437 | ||
DC399934.1 | - | 2, 3 | 212 |