Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001671.3 | NP_001001671.3 | 17 | 2246 | ||
XM_011545507.2 | XP_011543809.2 | 18 | 2473 | |||
XM_011545508.2 | XP_011543810.2 | 18 | 2473 | |||
XM_011545510.2 | XP_011543812.1 | 11 | 2986 | |||
XM_011545511.2 | XP_011543813.1 | 8 | 1497 | |||
XM_017029511.1 | XP_016885000.1 | 18 | 2475 | |||
GenBank mRNA | AK131412.1 | - | 14 | 1574 | ||
AK131477.1 | - | 18 | 2306 |