Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178137.1 | NP_001171608.1 | 4 | 1256 | ||
NM_014912.4 | NP_055727.3 | 4 | 1407 | |||
XM_005269630.4 | XP_005269687.1 | 4 | 1338 | |||
XM_006717714.3 | XP_006717777.1 | 4 | 1325 | |||
XM_006717715.3 | XP_006717778.1 | 4 | 1407 | |||
XM_011539514.2 | XP_011537816.1 | 4 | 1407 | |||
XM_011539515.2 | XP_011537817.1 | 4 | 1338 | |||
XM_011539518.2 | XP_011537820.1 | 4 | 1407 | |||
XM_011539519.2 | XP_011537821.1 | 4 | 1407 | |||
XM_011539520.2 | XP_011537822.1 | 4 | 1338 | |||
XM_011539522.2 | XP_011537824.1 | 4 | 1407 | |||
XM_011539523.2 | XP_011537825.1 | 4 | 1338 | |||
GenBank mRNA | AB023157.1 | - | 4 | 1223 | ||
AK127060.1 | - | 3 | 328 | |||
AK302576.1 | - | 4 | 1325 | |||
BC036444.2 | - | 4 | 1376 | 4838004 |