Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294468.1 | - | 2 | 132 | ||
AK316022.1 | - | 2 | 181 | |||
AK316283.1 | - | 2 | 191 | |||
BC130577.1 | - | 2 | 157 | 40146608 | ||
BC130579.1 | - | 2 | 157 | 40146610 | ||
CR749240.1 | - | 2 | 169 |