Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 3 | 1218 | ||
NM_001291447.1 | NP_001278376.1 | 3 | 1198 | |||
NM_005407.2 | NP_005398.2 | 2 | 1623 | |||
XM_011537064.1 | XP_011535366.1 | 2 | 1549 | |||
XM_011537065.1 | XP_011535367.1 | 2 | 1695 | |||
GenBank mRNA | AB002358.2 | - | 2 | 1610 | ||
AK293574.1 | - | 3 | 904 | |||
AK294051.1 | - | 2, 3 | 1404 | |||
AK296414.1 | - | 3 | 1017 | |||
AK299931.1 | - | 2 | 1497 | |||
BC136528.1 | - | 2 | 1358 | 9020518 | ||
BC136529.1 | - | 2 | 1358 | 9020519 | ||
X98834.1 | - | 2 | 1510 |