Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015793.2 | NP_056608.1 | 1373 | |||
GenBank mRNA | AK087709.1 | - | 1373 | |||
AK135682.1 | - | 1369 | ||||
AK166611.1 | - | 1211 | ||||
AK166612.1 | - | 1211 | ||||
BC099372.1 | - | 1198 | 30938910 | |||
BC138018.1 | - | 1367 | 8861037 | |||
BC145098.1 | - | 1204 | 9053646 |
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177420.1 | NP_001170891.1 | 10 | 1373 | ||
GenBank mRNA | BC084592.1 | - | 10 | 1363 |