Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007227.3 | NP_009158.3 | 1 | 1364 | ||
GenBank mRNA | U92642.1 | - | 1 | 1416 |
Set Membership: |
G-protein coupled receptor |