Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170433.1 | NP_001163904.1 | 28 | 3857 | ||
NM_026221.2 | NP_080497.1 | 27 | 3824 | |||
GenBank mRNA | AK003318.1 | - | 1 | 445 | ||
AK014559.1 | - | 11 | 1990 | |||
AK044496.1 | - | 18 | 3788 | |||
AK146575.1 | - | 27 | 3808 | |||
AK154482.1 | - | 27 | 3770 | |||
AK173133.1 | - | 28 | 3763 | |||
BC035209.1 | - | 27 | 3671 | |||
BC049862.1 | - | 6 | 1319 | |||
BC058176.1 | - | 15 | 2361 | |||
BC060078.1 | - | 18 | 4097 | |||
BC071257.1 | - | 8 | 1533 | |||
BC072603.1 | - | 27 | 3801 | 6830850 |