Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026964.3 | NP_081240.1 | 3 | 279 | ||
GenBank mRNA | AK007589.1 | - | 3 | 280 | ||
AK088037.1 | - | 3 | 290 | |||
BC049678.1 | - | 3 | 279 | 6595832 | ||
BC050938.1 | - | 2 | 257 | 6819562 |