Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027166.5 | NP_081442.1 | 3 | 1285 | ||
GenBank mRNA | AK046281.1 | - | 3 | 1285 | ||
AK049096.1 | - | 3 | 1286 | |||
AK049644.1 | - | 3 | 1284 | |||
AK137446.1 | - | 3 | 1257 | |||
AK154461.1 | - | 3 | 1323 | |||
AK167998.1 | - | 3 | 1303 | |||
AK169241.1 | - | 3 | 1288 | |||
BC085109.1 | - | 3 | 1305 | 30737658 |