Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027740.6 | NP_082016.1 | 5 | 516 | ||
GenBank mRNA | AK016991.1 | - | 4 | 440 | ||
AK030180.1 | - | 4 | 410 | |||
AK045302.1 | - | 5 | 495 | |||
AK050233.1 | - | 5 | 585 | |||
AK090075.1 | - | 5 | 519 | |||
BC050888.1 | - | 4 | 514 | |||
BC099926.1 | - | 5 | 603 | |||
BC144819.1 | - | 5 | 605 | |||
BC145801.1 | - | 5 | 605 | 40131104 |