Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282550.1 | NP_001269479.1 | 2 | 75 | ||
NM_001282551.1 | NP_001269480.1 | 2 | 171 | |||
NM_001282552.2 | NP_001269481.1 | 2 | 256 | |||
NM_001282554.1 | NP_001269483.1 | 2 | 171 | |||
NM_014962.3 | NP_055777.1 | 1 | 493 | |||
NM_181443.3 | NP_852108.2 | 2 | 256 | |||
XM_017027728.1 | XP_016883217.1 | 3 | 404 | |||
XM_017027729.1 | XP_016883218.1 | 3 | 336 | |||
XM_017027730.1 | XP_016883219.1 | 1 | 515 | |||
GenBank mRNA | AB023169.1 | - | 1 | 441 | ||
AK299281.1 | - | 2 | 256 | |||
AK316256.1 | - | 2 | 147 | |||
BC109315.1 | - | 2 | 104 | 40036410 | ||
BC109316.1 | - | 2 | 104 | 40036411 | ||
CB961606.1 | - | 2 | 177 | |||
DC314595.1 | - | 2 | 171 | |||
DC418438.1 | - | 2 | 75 | |||
DR759697.1 | - | 2 | 111 |