Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212531.2 | NP_997696.1 | 20 | 1706 | ||
GenBank mRNA | BC091227.1 | - | 20 | 1634 | 7317323 |