Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 3 | 359 | ||
NR_027410.1 | 11 | 2607 | ||||
XM_011521596.1 | XP_011519898.1 | 10 | 1204 | |||
XM_011521597.1 | XP_011519899.1 | 9 | 1155 | |||
XM_017022210.1 | XP_016877699.1 | 11 | 1508 | |||
XM_017022211.1 | XP_016877700.1 | 11 | 1277 | |||
XM_017022212.1 | XP_016877701.1 | 10 | 1831 | |||
XM_017022213.1 | XP_016877702.1 | 10 | 1233 | |||
XM_017022214.1 | XP_016877703.1 | 11 | 1508 | |||
XM_017022215.1 | XP_016877704.1 | 11 | 2030 | |||
XM_017022216.1 | XP_016877705.1 | 10 | 2628 | |||
XM_017022217.1 | XP_016877706.1 | 9 | 2550 | |||
XM_017022218.1 | XP_016877707.1 | 11 | 2657 | |||
GenBank mRNA | AK097551.1 | - | 3 | 348 | ||
BC143254.1 | - | 3 | 305 | 9051756 | ||
BC144610.1 | - | 1 | 96 | 9053141 | ||
BX647673.1 | - | 10 | 2576 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 3 | 359 | ||
NR_027409.1 | 11 | 2621 | ||||
XM_006720440.2 | XP_006720503.1 | 3 | 327 | |||
XM_017022018.1 | XP_016877507.1 | 9 | 2402 | |||
XM_017022019.1 | XP_016877508.1 | 8 | 2098 | |||
XM_017022020.1 | XP_016877509.1 | 1 | 449 | |||
GenBank mRNA | AB020662.1 | - | 2 | 339 | ||
AF163441.1 | - | 1 | 783 | |||
AF204231.1 | - | 2 | 676 | |||
AK123221.1 | - | 7 | 1965 | |||
BC032774.1 | - | 3 | 261 | |||
BC093673.1 | - | 3 | 305 | 7939518 | ||
BC104800.1 | - | 3 | 305 | 8143803 | ||
BC144609.1 | - | 1 | 96 | 9053140 | ||
BC150329.1 | - | 2 | 350 | |||
BC152410.1 | - | 2 | 339 | |||
BX648160.1 | - | 10 | 2575 | |||
BX648575.1 | - | 10 | 2328 | |||
CR749361.1 | - | 3 | 1512 |