Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304467.1 | NP_001291396.1 | 7 | 1015 | ||
NM_001304468.1 | NP_001291397.1 | 6 | 756 | |||
NM_212554.3 | NP_997719.2 | 7 | 1011 | |||
GenBank mRNA | AF318345.1 | - | 6 | 686 | ||
AK022354.1 | - | 3 | 462 | |||
AL832292.1 | - | 7 | 912 | |||
BC026167.1 | - | 7 | 946 | 4823003 |