Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015044.4 | NP_055859.1 | 2 | 214 | ||
XM_017023074.1 | XP_016878563.1 | 3 | 3903 | |||
XM_017023075.1 | XP_016878564.1 | 2 | 212 | |||
GenBank mRNA | AF165531.1 | - | 2 | 152 | ||
AF190863.1 | - | 2 | 158 | |||
AF233522.1 | - | 2 | 157 | |||
AF323754.1 | - | 2 | 156 | |||
AK024619.1 | - | 2 | 214 | |||
AK308425.1 | - | 2 | 201 | |||
BC000284.1 | - | 2 | 196 | 3356490 | ||
CN423115.1 | - | 2 | 211 | |||
DA439738.1 | - | 2 | 214 |