Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017107.2 | NP_058803.2 | 19 | 2475 | ||
XM_006257056.1 | XP_006257118.1 | 18 | 2610 | |||
XM_006257057.2 | XP_006257119.1 | 19 | 2552 | |||
XM_006257058.1 | XP_006257120.1 | 18 | 2580 |