Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175247.3 | NP_780456.2 | 5 | 709 | ||
GenBank mRNA | AK028202.1 | - | 5 | 682 | ||
AK048530.1 | - | 5 | 701 | |||
AK173170.1 | - | 5 | 674 | |||
AY528713.1 | - | 5 | 615 | |||
BC062116.1 | - | 5 | 677 | 6817400 |