Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030886.2 | NP_112148.2 | 19 | 3750 | ||
NM_198010.2 | NP_932127.2 | 18 | 2997 | |||
GenBank mRNA | AF130371.1 | - | 17 | 2538 | ||
AK160263.1 | - | 4 | 514 | |||
BC080807.1 | - | 9 | 1020 | |||
BC085166.1 | - | 18 | 2515 | |||
CB575216.1 | - | 5 | 624 |