Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198608.2 | NP_941010.2 | 22 | 3108 | ||
GenBank mRNA | AK146958.1 | - | 22 | 3109 | ||
AK167771.1 | - | 22 | 3103 | |||
AK173139.1 | - | 18 | 2382 | |||
BC059889.1 | - | 11 | 2715 | |||
BC079844.1 | - | 22 | 3080 | |||
BC110699.1 | - | 11 | 2715 |