Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004537.5 | NP_004528.1 | 15 | 3741 | ||
NM_139207.3 | NP_631946.1 | 16 | 3067 | |||
XM_006719425.3 | XP_006719488.1 | 14 | 2530 | |||
XM_011538390.2 | XP_011536692.2 | 16 | 2698 | |||
XM_011538393.2 | XP_011536695.1 | 16 | 2979 | |||
XM_017019338.1 | XP_016874827.1 | 16 | 2951 | |||
XM_017019339.1 | XP_016874828.1 | 14 | 2790 | |||
XM_017019340.1 | XP_016874829.1 | 14 | 2502 | |||
GenBank mRNA | AF086080.1 | - | 1 | 560 | ||
AF300335.2 | - | 1 | 893 | |||
AL162068.1 | - | 15 | 3380 | |||
DA137494.1 | - | 1 | 272 |