Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013703.3 | NP_001013725.2 | 6 | 694 | ||
XM_005254392.2 | XP_005254449.1 | 6 | 717 | |||
XM_011521599.2 | XP_011519901.1 | 6 | 717 | |||
XM_011521600.2 | XP_011519902.1 | 6 | 717 | |||
XM_017022219.1 | XP_016877708.1 | 6 | 717 | |||
GenBank mRNA | AB037759.1 | - | 2 | 160 | ||
AL832907.1 | - | 1 | 38 | |||
BC078179.1 | - | 6 | 672 |