Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015140.3 | NP_055955.1 | 3 | 574 | ||
GenBank mRNA | AL590888.1 | - | 3 | 512 | ||
BC001070.2 | - | 3 | 538 | 3504490 | ||
BX411143.2 | - | 3 | 572 | |||
CT841516.1 | - | 3 | 515 | |||
D63487.1 | - | 3 | 496 |