Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 30 | 6738 | ||
XM_005246237.2 | XP_005246294.1 | 32 | 6466 | |||
XM_005246239.2 | XP_005246296.1 | 32 | 6281 | |||
XM_005246240.2 | XP_005246297.1 | 25 | 4483 | |||
XM_005246241.1 | XP_005246298.1 | 25 | 4377 | |||
XM_005246242.4 | XP_005246299.1 | 25 | 4502 | |||
XM_006712189.3 | XP_006712252.1 | 31 | 6471 | |||
XM_006712193.3 | XP_006712256.1 | 25 | 7602 | |||
XM_011510479.2 | XP_011508781.1 | 31 | 6904 | |||
XM_011510483.2 | XP_011508785.2 | 31 | 6501 | |||
XM_017003157.1 | XP_016858646.1 | 31 | 6501 | |||
XM_017003158.1 | XP_016858647.1 | 27 | 6077 | |||
XM_017003159.1 | XP_016858648.1 | 28 | 6260 | |||
XM_017003160.1 | XP_016858649.1 | 15 | 2673 | |||
GenBank mRNA | AB037718.1 | - | 21 | 3795 | ||
AY603755.1 | - | 29 | 6351 |
Set Membership: |
G-protein coupled receptor |