Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABLES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031215.2 | NP_112492.2 | 5, 6 | 723 | ||
GenBank mRNA | BC003122.1 | - | 3, 4 | 258 | ||
BU166894.1 | - | 5, 6 | 436 |