Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007793.2 | NP_001007794.1 | 7 | 1036 | ||
NM_004725.3 | NP_004716.1 | 7 | 1036 | |||
GenBank mRNA | AF047472.1 | - | 7 | 864 | ||
AF047473.1 | - | 7 | 864 | |||
AF053304.1 | - | 7 | 832 | |||
AF081496.1 | - | 7 | 881 | |||
AK226060.1 | - | 2619 | ||||
AK293255.1 | - | 6 | 692 | |||
AK312546.1 | - | 7 | 904 | |||
BC005138.2 | - | 7 | 865 | 3504520 | ||
BC022438.1 | - | 7 | 857 | 4281970 | ||
BX647392.1 | - | 7 | 913 |