Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 18 | 2513 | ||
NM_001162426.1 | NP_001155898.1 | 18 | 2510 | |||
NM_001162427.1 | NP_001155899.1 | 17 | 2360 | |||
XM_005272211.1 | XP_005272268.1 | 18 | 2558 | |||
XM_006717271.1 | XP_006717334.1 | 18 | 2493 | |||
XM_011518979.2 | XP_011517281.1 | 18 | 2507 | |||
XM_017015096.1 | XP_016870585.1 | 19 | 2972 | |||
XM_017015097.1 | XP_016870586.1 | 19 | 2943 | |||
XM_017015098.1 | XP_016870587.1 | 18 | 2555 | |||
XM_017015099.1 | XP_016870588.1 | 17 | 2651 | |||
XM_017015100.1 | XP_016870589.1 | 17 | 2622 | |||
XM_017015101.1 | XP_016870590.1 | 17 | 2649 | |||
GenBank mRNA | AF013168.1 | - | 18 | 2500 | ||
AK303030.1 | - | 17 | 2343 | |||
AK308412.1 | - | 18 | 2494 | |||
D87683.1 | - | 5 | 884 | |||
KJ534953.1 | - | 16 | 2279 |