Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286839.1 | NP_001273768.1 | 3 | 499 | ||
NM_005766.3 | NP_005757.1 | 3 | 678 | |||
XM_011521046.2 | XP_011519348.1 | 3 | 678 | |||
XM_017020312.1 | XP_016875801.1 | 3 | 422 | |||
XM_017020313.1 | XP_016875802.1 | 2 | 1415 | |||
GenBank mRNA | AB008430.1 | - | 3 | 308 | ||
BC041595.1 | - | 3 | 525 | 5443230 | ||
BC071592.1 | - | 3 | 499 | 30342239 |