Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301132.1 | NP_001288061.1 | 2 | 1096 | ||
NM_014952.4 | NP_055767.3 | 2 | 969 | |||
XM_011521366.2 | XP_011519668.2 | 2 | 1093 | |||
XM_011521367.2 | XP_011519669.2 | 2 | 1098 | |||
XM_011521368.2 | XP_011519670.2 | 2 | 1092 | |||
XM_011521369.2 | XP_011519671.1 | 2 | 947 | |||
GenBank mRNA | AB023162.1 | - | 2 | 1096 | ||
AL833923.1 | - | 1 | 61 | |||
BC022782.2 | - | 2 | 908 | 3903581 |