Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014964.4 | NP_055779.2 | 3 | 861 | ||
NM_148921.3 | NP_683723.2 | 3 | 861 | |||
GenBank mRNA | AB028988.1 | - | 3 | 826 | ||
AF062084.1 | - | 1 | 413 | |||
AF062085.1 | - | 1 | 413 | |||
AK001491.1 | - | 1 | 554 | |||
AK001996.1 | - | 1 | 580 | |||
AK024115.1 | - | 3 | 841 | |||
AK298006.1 | - | 2 | 699 | |||
BC093972.1 | - | 1 | 486 | 7939817 | ||
BC093974.1 | - | 1 | 486 | 7939819 | ||
BC143264.1 | - | 1 | 487 | 9051766 |