Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286752.1 | NP_001273681.1 | 10 | 1687 | ||
NM_017438.4 | NP_059134.1 | 9 | 1544 | |||
NR_040087.2 | 8 | 1455 | ||||
XM_011529636.1 | XP_011527938.1 | 9 | 1593 | |||
XM_011529637.1 | XP_011527939.1 | 9 | 1658 | |||
XM_011529638.2 | XP_011527940.1 | 9 | 1566 | |||
XM_011529639.1 | XP_011527941.1 | 9 | 1344 | |||
XM_011529640.2 | XP_011527942.1 | 9 | 1609 | |||
XM_011529642.1 | XP_011527944.1 | 10 | 1687 | |||
XM_011529643.1 | XP_011527945.1 | 10 | 1638 | |||
XM_011529644.1 | XP_011527946.1 | 10 | 1199 | |||
XM_017028403.1 | XP_016883892.1 | 9 | 1135 | |||
XM_017028404.1 | XP_016883893.1 | 10 | 1243 | |||
GenBank mRNA | AF391112.1 | - | 8 | 1022 | ||
AK001660.1 | - | 9 | 1111 | |||
AK095215.1 | - | 8 | 2411 | |||
AK126368.1 | - | 2 | 1621 | |||
BC036556.2 | - | 10 | 1361 | 5266627 |