Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 10, 11 | 1447 | ||
NM_001008212.1 | NP_001008213.1 | 9, 10 | 1378 | |||
NM_001008213.1 | NP_001008214.1 | 10, 11 | 1432 | |||
NM_021980.4 | NP_068815.2 | 8, 9 | 1226 | |||
GenBank mRNA | AF061034.1 | - | 10, 11 | 1320 | ||
AF420371.1 | - | 10, 11 | 1304 | |||
AF420372.1 | - | 8, 9 | 1083 | |||
AF420373.1 | - | 9, 10 | 1235 | |||
AK055403.1 | - | 9, 10 | 1667 | |||
AK122746.1 | - | 1, 2 | 741 | |||
AK303715.1 | - | 8, 9 | 1244 | |||
AK316171.1 | - | 8, 9 | 1266 | |||
BC013876.2 | - | 8, 9 | 1139 | 3831267 | ||
BC032762.1 | - | 10, 11 | 1334 | 3457195 | ||
KU178502.1 | - | 7, 8 | 994 |