Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 15 | 2859 | ||
XM_005244731.3 | XP_005244788.1 | 15 | 2964 | |||
GenBank mRNA | AB209210.1 | - | 9 | 4343 | ||
AF006011.1 | - | 15 | 2815 | |||
AK093189.1 | - | 8 | 2099 | |||
AK095867.1 | - | 8 | 2077 | |||
BC017225.2 | - | 13 | 2584 | |||
BC050454.1 | - | 9 | 2204 | 6201003 |