Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 10 | 3049 | ||
NM_001006642.3 | NP_001006643.1 | 10 | 3294 | |||
NM_001265614.2 | NP_001252543.1 | 11 | 3330 | |||
NM_052901.4 | NP_443133.2 | 10 | 2912 | |||
NR_049766.2 | 10 | 2955 | ||||
XM_005251688.1 | XP_005251745.1 | 11 | 3085 | |||
XM_005251689.4 | XP_005251746.1 | 10 | 3308 | |||
XM_006716948.2 | XP_006717011.1 | 11 | 3486 | |||
XM_006716949.3 | XP_006717012.1 | 11 | 3346 | |||
GenBank mRNA | AB067483.1 | - | 11 | 3329 | ||
AJ619963.1 | - | 10 | 3213 | |||
AJ619989.1 | - | 10 | 2837 | |||
AJ619990.1 | - | 10 | 2932 | |||
AJ619991.1 | - | 10 | 2894 | |||
AJ619992.1 | - | 10 | 2880 | |||
AY358515.1 | - | 10 | 2887 | |||
BC005163.2 | - | 6 | 2346 | |||
BC044631.1 | - | 1 | 1472 | |||
BC061925.1 | - | 1 | 469 | |||
BC089448.1 | - | 10 | 2903 | 30558551 |