Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016329.3 | NP_057413.2 | 14, 15 | 1870 | ||
XM_005265221.3 | XP_005265278.1 | 14, 15 | 1986 | |||
XM_006713203.3 | XP_006713266.1 | 15, 16 | 2108 | |||
XM_006713204.3 | XP_006713267.1 | 14, 15 | 1808 | |||
XM_011533824.2 | XP_011532126.1 | 15, 16 | 1984 | |||
XM_011533825.2 | XP_011532127.1 | 15, 16 | 1831 | |||
GenBank mRNA | AB189472.1 | - | 13, 14 | 1494 | ||
AF168132.1 | - | 14, 15 | 1807 | |||
AK313965.1 | - | 14, 15 | 1865 | |||
AL080140.1 | - | 11, 12 | 1148 | |||
BC014614.2 | - | 14, 15 | 1754 | 3029598 |