Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171905.1 | NP_001165376.1 | 4 | 1357 | ||
NM_152395.2 | NP_689608.2 | 3 | 1531 | |||
NR_033268.1 | 2 | 1618 | ||||
GenBank mRNA | AK055827.1 | - | 3 | 1433 |