Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003823.3 | NP_003814.1 | 1 | 365 | ||
GenBank mRNA | BC034349.1 | - | 1 | 356 | 4752507 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 36 | 4982 |