Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178833.5 | NP_849155.2 | 4 | 1004 | ||
XM_006714085.2 | XP_006714148.1 | 4 | 1003 | |||
GenBank mRNA | AK172823.1 | - | 4 | 1004 | ||
BC009732.1 | - | 1 | 72 | |||
BC047447.1 | - | 4 | 534 | 5295722 |