Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184745.1 | NP_001171674.1 | 4 | 1044 | ||
NM_033215.4 | NP_149992.3 | 4 | 1987 | |||
XM_005272687.3 | XP_005272744.1 | 4 | 2005 | |||
XM_017029932.1 | XP_016885421.1 | 4 | 6415 | |||
XM_017029933.1 | XP_016885422.1 | 4 | 6412 | |||
GenBank mRNA | AK056297.1 | - | 3 | 1404 | ||
AK056909.1 | - | 4 | 1812 | |||
AK127066.1 | - | 1 | 2645 | |||
AK303738.1 | - | 5 | 1310 | |||
BC110405.1 | - | 1 | 572 | |||
BC131588.1 | - | 4 | 1017 |