Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 2 | 344 | ||
XM_011514422.1 | XP_011512724.1 | 2 | 353 | |||
XM_011514423.2 | XP_011512725.1 | 3 | 455 | |||
XM_011514424.1 | XP_011512726.1 | 2 | 353 | |||
XM_011514425.1 | XP_011512727.1 | 2 | 353 | |||
XM_011514426.1 | XP_011512728.1 | 2 | 353 | |||
XM_011514428.1 | XP_011512730.1 | 2 | 353 | |||
XM_011514430.1 | XP_011512732.1 | 2 | 353 | |||
XM_017010589.1 | XP_016866078.1 | 2 | 1064 | |||
GenBank mRNA | AJ318215.1 | - | 1 | 269 | ||
AK125228.1 | - | 2 | 345 | |||
AY145132.1 | - | 1 | 269 | |||
BC028159.1 | - | 2 | 319 | |||
CR749511.1 | - | 2 | 344 | |||
CR749841.1 | - | 2 | 343 |